Learn More
Aviva Systems Biology Corp POU4F1 Antibody - C-terminal region (ARP38763_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38763_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POU4F1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human POU4F1. Alternate names: BRN3A, RDC-1, Oct-T1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53113