Learn More
Aviva Systems Biology Corp POU4F2 Antibody - N-terminal region (ARP38501_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38501_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POU4F2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POU4F2. Alternate names: BRN3.2, BRN3B, Brn-3bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53107