Learn More
Aviva Systems Biology Corp POU4F3 Antibody - middle region (ARP33065_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33065_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POU4F3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human POU4F3. Alternate names: BRN3C, DFNA15ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54867