Learn More
Aviva Systems Biology Corp POU5F1 Antibody - N-terminal region (P100956_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100956_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POU5F1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POU5F1. Alternate names: OCT3, OCT4, OTF3, OTF4, OTF-3, Oct-3, Oct-4ApplicationWB. Species reactivity: Dog, Goat, Human, Pig, Rabbit
Catalog No. 50-014-62798