Learn More
Aviva Systems Biology Corp POU5F1 Antibody - middle region (ARP40085_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40085_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POU5F1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human POU5F1. Alternate names: MGC22487, OCT3, OCT4, OTF3, OTF4, OTF-3, Oct-3, Oct-4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-57289