Learn More
Aviva Systems Biology Corp POU6F2 Antibody - C-terminal region (ARP38929_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38929_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POU6F2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human POU6F2. Alternate names: RPF-1, WT5, WTSLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-766