Learn More
Aviva Systems Biology Corp POU6F2 Antibody - N-terminal region (ARP31534_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31534_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POU6F2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POU6F2. Alternate names: RPF-1, WT5, WTSLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54258