Learn More
Med Vet International BioHazard Bag, J0724, 100/Pack
Catalog No. 5000495374
Encompass_Preferred
Handy size bag and holder are ideal for collecting lab samples like blood tubes. Bag stays open until you are ready to dispose of it. These 1.5mm thick polypropylene bags can be safely autoclaved. An indicator patch darkens when sterilized. Very inexpensive. 3.6" x 5.1" x 8.4." 100/pk.
Catalog No. 50-004-95374 Supplier Med Vet International Supplier No. JOR724
Handy size bag and holder are ideal for collecting lab samples like blood tubes. Bag stays open until you are ready to dispose of it. These 1.5mm thick polypropylene bags can be safely autoclaved. An indicator patch darkens when sterilized. Very inexpensive. 3.6" x 5.1" x 8.4." 100/pk.