Learn More
Cole-Parmer Oakton Conductivity Calibration Pouches, 84 uS; 20/Pk
Supplier:  Cole-Parmer 3565308
Encompass_Preferred
Single-use 20-mL pouches are sealed to ensure freshness—take these convenient standard solutions anywhere! Don't worry about transferring solution into another container—calibrate directly from the pouch. A µS-to-ppm conversion table is printed on each pack for quick reference.
Catalog No. 50-196-1364