Learn More
Med Vet International McKesson Sterilization Pouch, 12 x 15 Inch, 16-6422, 1000/Case
Catalog No. 503042844
Encompass_Preferred
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 12 X 15 Inch Transparent Blue / White Self Seal Paper / Film For use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 12 x 15 inches
Catalog No. 50-304-2844 Supplier Med Vet International Supplier No. 960947CS
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 12 X 15 Inch Transparent Blue / White Self Seal Paper / Film For use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 12 x 15 inches