Learn More
Med Vet International McKesson Sterilization Pouch, 12 x 18 Inch, 16-6421, 200/Box
Catalog No. 503043888
Encompass_Preferred
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 12 X 18 Inch Transparent Blue / White Self Seal Paper / Film For use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 12 x 18 inches
Catalog No. 50-304-3888 Supplier Med Vet International Supplier No. 960948BX
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 12 X 18 Inch Transparent Blue / White Self Seal Paper / Film For use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 12 x 18 inches