Learn More
Med Vet International McKesson Argent Sure-Check Sterilization Pouch, 3½ x 9 Inch, 73-SSP381, 200/Box
Catalog No. 503045059
Encompass_Preferred
Sterilization Pouch McKesson Argent™ Sure-Check Ethylene Oxide (EO) Gas / Steam 3-1/2 X 9 Inch Transparent / Blue Self Seal Paper / Film Class 4 indicator on outside and inside of the pouch, compatible with steam and ethylene oxide Triple seal (heat sealed) secured edges to help prevent separation Secure Seal -- Extra wide chevron seal to help prevent instrument breaching Medical grade paper Clear transparent film to view instrument Thumb notch makes pouch easy to open and grip 3½ x 9 inches
Catalog No. 50-304-5059 Supplier Med Vet International Supplier No. 527134BX
Sterilization Pouch McKesson Argent™ Sure-Check Ethylene Oxide (EO) Gas / Steam 3-1/2 X 9 Inch Transparent / Blue Self Seal Paper / Film Class 4 indicator on outside and inside of the pouch, compatible with steam and ethylene oxide Triple seal (heat sealed) secured edges to help prevent separation Secure Seal -- Extra wide chevron seal to help prevent instrument breaching Medical grade paper Clear transparent film to view instrument Thumb notch makes pouch easy to open and grip 3½ x 9 inches