Learn More
Med Vet International McKesson Sterilization Pouch, 3½ x 5 Inch, 16-6420, 200/Box
Catalog No. 503045189
Encompass_Preferred
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 3-1/2 X 5 Inch Transparent Blue / White Self Seal Paper / Film For use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 3½ x 5 inches
Catalog No. 50-304-5189 Supplier Med Vet International Supplier No. 960942BX
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 3-1/2 X 5 Inch Transparent Blue / White Self Seal Paper / Film For use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 3½ x 5 inches