Learn More
Med Vet International McKesson Sterilization Pouch, 3½ x 9 Inch, 16-6423, 200/Box
Catalog No. 503045190
Encompass_Preferred
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 3-1/2 X 9 Inch Transparent Blue / White Self Seal Paper / Film Self-sealing, which makes them easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 3½ x 9 inches
Catalog No. 50-304-5190 Supplier Med Vet International Supplier No. 960943BX
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 3-1/2 X 9 Inch Transparent Blue / White Self Seal Paper / Film Self-sealing, which makes them easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 3½ x 9 inches