Learn More
Med Vet International McKesson Argent Sure-Check Sterilization Pouch, 5-1/4 x 10 Inch, 73-SSP382, 2000/Case
Catalog No. 503044384
Encompass_Preferred
Sterilization Pouch McKesson Argent™ Sure-Check Ethylene Oxide (EO) Gas / Steam 5-1/4 X 10 Inch Transparent / Blue Self Seal Paper / Film Class 4 indicator on outside and inside of the pouch, compatible with steam and ethylene oxide Triple seal (heat sealed) secured edges to help prevent separation Secure Seal -- Extra wide chevron seal to help prevent instrument breaching Medical grade paper Clear transparent film to view instrument Thumb notch makes pouch easy to open and grip 5¼ x 10 inches
Catalog No. 50-304-4384 Supplier Med Vet International Supplier No. 524880CS
Sterilization Pouch McKesson Argent™ Sure-Check Ethylene Oxide (EO) Gas / Steam 5-1/4 X 10 Inch Transparent / Blue Self Seal Paper / Film Class 4 indicator on outside and inside of the pouch, compatible with steam and ethylene oxide Triple seal (heat sealed) secured edges to help prevent separation Secure Seal -- Extra wide chevron seal to help prevent instrument breaching Medical grade paper Clear transparent film to view instrument Thumb notch makes pouch easy to open and grip 5¼ x 10 inches