Learn More
Med Vet International McKesson Sterilization Pouch, 5¼ x 10 Inch, 16-6424, 200/Box
Catalog No. 503045123
Encompass_Preferred
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 5-1/4 X 10 Inch Transparent Blue / White Self Seal Paper / Film Self-sealing, which makes them easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink
Catalog No. 50-304-5123 Supplier Med Vet International Supplier No. 960944BX
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 5-1/4 X 10 Inch Transparent Blue / White Self Seal Paper / Film Self-sealing, which makes them easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink