Learn More
Med Vet International McKesson Sterilization Pouch, 7½ x 13 Inch, 16-6425, 1000/Case
Catalog No. 503045380
Encompass_Preferred
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 7-1/2 X 13 Inch Transparent Blue / White Self Seal Paper / Film For use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 7½ x 13 inches
Catalog No. 50-304-5380 Supplier Med Vet International Supplier No. 960945CS
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 7-1/2 X 13 Inch Transparent Blue / White Self Seal Paper / Film For use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 7½ x 13 inches