Learn More
Med Vet International McKesson Sterilization Pouch, 7½ x 13 Inch, 16-6425, 200/Box
Catalog No. 503045379
Encompass_Preferred
For use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 7.5 x 13 inches
Catalog No. 50-304-5379 Supplier Med Vet International Supplier No. 960945BX
For use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 7.5 x 13 inches