Learn More
Med Vet International McKesson Argent Sure-Check Sterilization Pouch, 8 x 16 Inch, 73-SSP387, 200/Box
Catalog No. 503044480
Encompass_Preferred
Sterilization Pouch McKesson Argent™ Sure-Check Ethylene Oxide (EO) Gas / Steam 8 X 16 Inch Transparent / Blue Self Seal Paper / Film Class 4 indicator on outside and inside of the pouch, compatible with steam and ethylene oxide Triple seal (heat sealed) secured edges to help prevent separation Secure Seal -- Extra wide chevron seal to help prevent instrument breaching Medical grade paper Clear transparent film to view instrument Thumb notch makes pouch easy to open and grip Not made with natural rubber latex
Catalog No. 50-304-4480 Supplier Med Vet International Supplier No. 527135BX
Sterilization Pouch McKesson Argent™ Sure-Check Ethylene Oxide (EO) Gas / Steam 8 X 16 Inch Transparent / Blue Self Seal Paper / Film Class 4 indicator on outside and inside of the pouch, compatible with steam and ethylene oxide Triple seal (heat sealed) secured edges to help prevent separation Secure Seal -- Extra wide chevron seal to help prevent instrument breaching Medical grade paper Clear transparent film to view instrument Thumb notch makes pouch easy to open and grip Not made with natural rubber latex