Learn More
Med Vet International McKesson Sterilization Pouch, 8 x 16 Inch, 16-6426, 200/Box
Catalog No. 503045187
Encompass_Preferred
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 8 X 16 Inch Transparent Blue / White Self Seal Paper / Film Sterilization pouch for use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 8 x 16 inches
Catalog No. 50-304-5187 Supplier Med Vet International Supplier No. 960946BX
Sterilization Pouch McKesson Ethylene Oxide (EO) Gas / Steam 8 X 16 Inch Transparent Blue / White Self Seal Paper / Film Sterilization pouch for use with steam and ethylene oxide (EO) sterilizers Self-sealing, which makes pouches easy to open and close No autoclave tape necessary Internal and external chemical indicators Lead free ink 8 x 16 inches