Learn More
Med Vet International 7 1/2inx13in Self-Sealing Sterilization Pouches, 200/bx
Catalog No. 502522536
Encompass_Preferred
Safer sterilization with a complete and uniform seal of the pouch. Quicker and extremely easy to make a perfect seal every time as compared to other competitive pouches. New pre-folded flap closure folds perfectly into place, eliminating time consuming flap adjustment and helping to ensure the best seal possible. This helps eliminate misaligned seals that can compromise sterility. Reinforced plastic pouch and rugged medical-grade paper guard against tears and perforations. Pouches are constructed with dual internal and external indicators that change color when the optimum sterilization temperature has been reached, both in the autoclave chamber and inside the pouch.
Catalog No. 50-252-2536 Supplier Med Vet International Supplier No. 442103
Safer sterilization with a complete and uniform seal of the pouch. Quicker and extremely easy to make a perfect seal every time as compared to other competitive pouches. New pre-folded flap closure folds perfectly into place, eliminating time consuming flap adjustment and helping to ensure the best seal possible. This helps eliminate misaligned seals that can compromise sterility. Reinforced plastic pouch and rugged medical-grade paper guard against tears and perforations. Pouches are constructed with dual internal and external indicators that change color when the optimum sterilization temperature has been reached, both in the autoclave chamber and inside the pouch.