Learn More
Research Products International Corp Pour Boat Weigh Dish 137 ml
SDP
Catalog No. 502979092
Encompass
Flexible polystyrene boats feature smooth surfaces to provide accurate pouring with minimal sample loss. Disposable, stackable dishes are biologically inert, contaminant-free, and resistant to most dilute acids, alcohols, bases, and aqueous solutions. Withstand temperatures -10°C to +70°C and can be used with microwave moisture analyzers. Food-grade compliant, white color.
Catalog No. 50-297-9092 Supplier Research Products International Corp Supplier No. 120321
Flexible polystyrene boats feature smooth surfaces to provide accurate pouring with minimal sample loss. Disposable, stackable dishes are biologically inert, contaminant-free, and resistant to most dilute acids, alcohols, bases, and aqueous solutions. Withstand temperatures -10°C to +70°C and can be used with microwave moisture analyzers. Food-grade compliant, white color.