Learn More
Research Products International Corp Pour Boat Weigh Dish
SDP
Catalog No. 502979093
Encompass
Flexible polystyrene boats feature smooth surfaces to provide accurate pouring with minimal sample loss. Disposable, stackable dishes are biologically inert, contaminant-free, and resistant to most dilute acids, alcohols, bases, and aqueous solutions. Withstand temperatures -10°C to +70°C and can be used with microwave moisture analyzers. Food-grade compliant, white color.
Catalog No. 50-297-9093 Supplier Research Products International Corp Supplier No. 120322
Flexible polystyrene boats feature smooth surfaces to provide accurate pouring with minimal sample loss. Disposable, stackable dishes are biologically inert, contaminant-free, and resistant to most dilute acids, alcohols, bases, and aqueous solutions. Withstand temperatures -10°C to +70°C and can be used with microwave moisture analyzers. Food-grade compliant, white color.