Learn More
Research Products International Corp Pour Boat Weigh Dsh AntiStatic
SDP
Supplier:  Research Products International Corp 120323
Encompass
Flexible polystyrene boats feature smooth surfaces to provide accurate pouring with minimal sample loss. Disposable, stackable dishes are biologically inert, contaminant-free, and resistant to most dilute acids, alcohols, bases, and aqueous solutions. Withstand temperatures -10°C to +70°C and can be used with microwave moisture analyzers. Food-grade compliant, white color.
Catalog No. 50-297-9094