Learn More
Analytical Sales & Services Pour-boat Weighing Dish - Medium
Catalog No. 5000806839
Encompass
Pour-boat Weighing Dishes - Medium • Pour-spout design easily dispenses samples without flexing the dish. • Flat-bottom dishes are made from 12-mil thick polystyrene for better strength, stability, and handling L x W x H: 83 x 132 x 26 mm Color: White
Catalog No. 50-008-06839 Supplier Analytical Sales & Services Supplier No. PBWM
Pour-boat Weighing Dishes - Medium • Pour-spout design easily dispenses samples without flexing the dish. • Flat-bottom dishes are made from 12-mil thick polystyrene for better strength, stability, and handling L x W x H: 83 x 132 x 26 mm Color: White