Learn More
Analytical Sales & Services Pour-boat Weighing Dish - Small
Catalog No. 5000806757
Encompass
Pour-boat Weighing Dishes - Small\n\n• Pour-spout design easily dispenses samples without flexing the dish\n• Flat-bottom dishes are made from 12-mil thick polystyrene for better strength, stability, and handling\n• Length: 43mm\n• Width: 58mm\n• Height: 13mm\n• Color: White\n\nQty/Pkg: 500
Catalog No. 50-008-06757 Supplier Analytical Sales & Services Supplier No. PBWS
Pour-boat Weighing Dishes - Small\n\n• Pour-spout design easily dispenses samples without flexing the dish\n• Flat-bottom dishes are made from 12-mil thick polystyrene for better strength, stability, and handling\n• Length: 43mm\n• Width: 58mm\n• Height: 13mm\n• Color: White\n\nQty/Pkg: 500