Learn More
Sigma Aldrich Fine Chemicals Biosciences Pour Boat Weighing Dishes
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HS120227
Encompass
flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Catalog No. 50-290-0467