Learn More
SPECTRUM QUALITY STANDARDS PP50-F [-50C Pour Point] [Fuel Balance] (8 oz)
Catalog No. NC0823416
Encompass_Preferred
This standard is made accordingly to ASTM D97 with a pour point approximate value of -45°C. This is not compatible with a phase instrument.
Catalog No. NC0823416 Supplier SPECTRUM QUALITY STANDARDS Supplier No. PP50F
This standard is made accordingly to ASTM D97 with a pour point approximate value of -45°C. This is not compatible with a phase instrument.