Learn More
Cole-Parmer Cole-Parmer Stainless steel graduated pouring beaker, 32 oz/1000 mL
Catalog No. NC9656045 Shop All Cole-Parmer Products
Encompass_Preferred
These graduated pouring beakers Features easy grip handles for convenient pouring. Beakers are graduated in mL and oz and constructed of medium gauge 304 stainless steel.
Catalog No. NC9656045 Supplier Cole-Parmer Supplier No. 0722200
These graduated pouring beakers Features easy grip handles for convenient pouring. Beakers are graduated in mL and oz and constructed of medium gauge 304 stainless steel.