Learn More
Microsolv Technology Corporation Pouring Ring, Replacement, Natural, for HPLC Mobile Phase Reservoir Bottles. GL32 Threads. 1 EA.
Catalog No. 502270190
Encompass_Preferred
Using this Natural color Polypropylene Pouring or "Drip Ring" can make decanting liquids from any bottle with a GL32 Thread easier and prevent that last drop from getting away from you. This ring will fit deeper into the mouth of the Bottle, making is more secure and less likely to fall out.
This item is not returnable. View return policy
Catalog No. 50-227-0190 Supplier Microsolv Technology Corporation Supplier No. 58807Z32
Using this Natural color Polypropylene Pouring or "Drip Ring" can make decanting liquids from any bottle with a GL32 Thread easier and prevent that last drop from getting away from you. This ring will fit deeper into the mouth of the Bottle, making is more secure and less likely to fall out.