Learn More
Chemglass Life Sciences Pouring Ring, GL-32, Clear, Polypropylene
SDP
Catalog No. 501215142
Encompass_Preferred
Steam autoclavable to 140 degreesC.All pouring rings are polypropylene except for the CLS-1482-30 which is made of high temperature ETFE (Ethylene Tetrafluoroethylene); max temp is 180 degreesC.
Catalog No. 50-121-5142 Supplier Chemglass Life Sciences Supplier No. CLS148212
Steam autoclavable to 140 degreesC.All pouring rings are polypropylene except for the CLS-1482-30 which is made of high temperature ETFE (Ethylene Tetrafluoroethylene); max temp is 180 degreesC.