Learn More
Chemglass Life Sciences Pouring Ring, GL-45, Green, Polypropylene
SDP
Catalog No. 501215144
Encompass_Preferred
Steam autoclavable to 140 degreesC.All pouring rings are polypropylene except for the CLS-1482-30 which is made of high temperature ETFE (Ethylene Tetrafluoroethylene); max temp is 180 degreesC.
Catalog No. 50-121-5144 Supplier Chemglass Life Sciences Supplier No. CLS148214
Steam autoclavable to 140 degreesC.All pouring rings are polypropylene except for the CLS-1482-30 which is made of high temperature ETFE (Ethylene Tetrafluoroethylene); max temp is 180 degreesC.