Learn More
Chemglass Life Sciences Pouring Ring, GL-45, Clear, Polypropylene
SDP
Catalog No. NC2044763
Encompass_Preferred
Steam autoclavable to 140 degreesC.All pouring rings are polypropylene except for the CLS-1482-30 which is made of high temperature ETFE (Ethylene Tetrafluoroethylene); max temp is 180 degreesC.
Catalog No. NC2044763 Supplier Chemglass Life Sciences Supplier No. CLS148217
Steam autoclavable to 140 degreesC.All pouring rings are polypropylene except for the CLS-1482-30 which is made of high temperature ETFE (Ethylene Tetrafluoroethylene); max temp is 180 degreesC.