Learn More
Selleck Chemical LLC Povidone iodine S4678-100mg
SDP
Catalog No. 503109165
Encompass_Preferred
Povidone iodine (Betadine PVP iodine PVP-I Isodine) is a stable chemical complex of polyvinylpyrrolidone (povidone PVP) and elemental iodine Povidone iodine displays excellent antibacterial activity against MRSA and MSSA strains with MICs of 31 25 mg/L and 7 82 mg/L respectively Povidone iodine is used as a topical antiseptic in surgery and for skin and mucous membrane infections as well as as an aerosol
Catalog No. 50-310-9165 Supplier Selleck Chemical LLC Supplier No. S4678100MG
Povidone iodine (Betadine PVP iodine PVP-I Isodine) is a stable chemical complex of polyvinylpyrrolidone (povidone PVP) and elemental iodine Povidone iodine displays excellent antibacterial activity against MRSA and MSSA strains with MICs of 31 25 mg/L and 7 82 mg/L respectively Povidone iodine is used as a topical antiseptic in surgery and for skin and mucous membrane infections as well as as an aerosol