Learn More
Med Vet International Povidone Iodine Solution 1 Gallon, each
Catalog No. 501180479
Encompass_Preferred
Prep solutions contain 10% weight by volume Povidone Iodine, USP solution equivalent to 1% available iodine, and are effective for skin and mucous membrane preparation prior to surgical proscrub. Solutions are antimicrobial with surfactants. Compared to Betadine.
Catalog No. 50-118-0479 Supplier Med Vet International Supplier No. PVPSOLG
Prep solutions contain 10% weight by volume Povidone Iodine, USP solution equivalent to 1% available iodine, and are effective for skin and mucous membrane preparation prior to surgical proscrub. Solutions are antimicrobial with surfactants. Compared to Betadine.