Learn More
Med Vet International Povidone Iodine Solution 16oz
Catalog No. 502522193
Encompass_Preferred
Prep solutions contain 10% weight by volume Povidone Iodine, USP solution equivalent to 1% available iodine, and are effective for skin and mucous membrane preparation prior to surgical proscrub solutions are antimicrobial solutions with surfactants, and contain 7.5% weight by volume Povidone Iodine, USP solution equivalent to 0.75% available iodine. Each is effective as a surgical scrub or first aid antiseptic. Compared to Betadine.
Catalog No. 50-252-2193 Supplier Med Vet International Supplier No. PVPSOLP
Prep solutions contain 10% weight by volume Povidone Iodine, USP solution equivalent to 1% available iodine, and are effective for skin and mucous membrane preparation prior to surgical proscrub solutions are antimicrobial solutions with surfactants, and contain 7.5% weight by volume Povidone Iodine, USP solution equivalent to 0.75% available iodine. Each is effective as a surgical scrub or first aid antiseptic. Compared to Betadine.