Learn More
Foxx Life Sciences OLGA™ Povitsky (Roux) Cell Culture Bottle with Offset Tooled Neck, 5L (5000mL), Borosilicate Glass, 1/EA
Catalog No. 502061599
Encompass_Preferred
Foxx Life Sciences in collaboration with BOROSIL has developed its own variation of the Povitsky bottle, the OLGA™ Povitsky Culture Bottle. Foxx named OLGA after designer name Dr. Olga R. Povitsky with the leading quality of 3.3 Borosilicate Type 1 Glass. This quality glassware is manufactured from USP Type 1, ASTM E-438/Type I/Class A borosilicate glass that scientists have come to trust. The OLGA™ Povitsky Culture Bottles are highly resistant to thermal shock, with an industry-leading maximum temperature rating of 142C. The OLGA™ bottles have rectangular cross-sections with an offset rimmed neck. These bottles can be stacked, reducing the incubator space needed. The OLGA™ Povitsky Culture Bottle was designed for the preparation of toxins, mold growth, and general tissue culture work. Safety, versatility, and ease of use can all be found in Foxx Life Sciences glass bottles.
Catalog No. 50-206-1599 Supplier Foxx Life Sciences Supplier No. 1296033FLS
Foxx Life Sciences in collaboration with BOROSIL has developed its own variation of the Povitsky bottle, the OLGA™ Povitsky Culture Bottle. Foxx named OLGA after designer name Dr. Olga R. Povitsky with the leading quality of 3.3 Borosilicate Type 1 Glass. This quality glassware is manufactured from USP Type 1, ASTM E-438/Type I/Class A borosilicate glass that scientists have come to trust. The OLGA™ Povitsky Culture Bottles are highly resistant to thermal shock, with an industry-leading maximum temperature rating of 142C. The OLGA™ bottles have rectangular cross-sections with an offset rimmed neck. These bottles can be stacked, reducing the incubator space needed. The OLGA™ Povitsky Culture Bottle was designed for the preparation of toxins, mold growth, and general tissue culture work. Safety, versatility, and ease of use can all be found in Foxx Life Sciences glass bottles.