Learn More
Medchemexpress LLC Povorcitinib (phosphate) | 1637677-33-8 | 98.8% | C23H25F5N7O5P | 25 MG
SDP
Catalog No. 5000355188
Encompass_Preferred
Povorcitinib phosphate is a potent and selective JAK1 inhibitor. It effectively reduces abscesses and inflammatory nodules and is utilized in studies of cutaneous lupus erythematosus (CLE) and lichen planus (LP). It exhibits anti-inflammatory activity by regulating the expression of TNF-α and TGF-β.
  • Potent and selective JAK1 inhibitor
  • Reduces abscesses and inflammatory nodules
  • Suitable for studying cutaneous lupus erythematosus (CLE)
  • Suitable for studying lichen planus (LP)
  • Regulates TNF-α and TGF-β expression
  • Solid appearance
  • White to off-white color
  • High purity (98.8%)
Catalog No. 50-003-55188 Supplier Medchemexpress LLC Supplier No. 25MGHY145588A
Povorcitinib phosphate is a potent and selective JAK1 inhibitor. It effectively reduces abscesses and inflammatory nodules and is utilized in studies of cutaneous lupus erythematosus (CLE) and lichen planus (LP). It exhibits anti-inflammatory activity by regulating the expression of TNF-α and TGF-β.
  • Potent and selective JAK1 inhibitor
  • Reduces abscesses and inflammatory nodules
  • Suitable for studying cutaneous lupus erythematosus (CLE)
  • Suitable for studying lichen planus (LP)
  • Regulates TNF-α and TGF-β expression
  • Solid appearance
  • White to off-white color
  • High purity (98.8%)