Learn More
TARGETMOL CHEMICALS INC POVORCITINIB PHOSPHATE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T39113L5MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Povorcitinib phosphate is a selective JAK1 inhibitor and can be used in studies about the treatment of cutaneous lupus erythematosus and Lichen planus. purity: 97%
Catalog No. 50-258-0068