Learn More
Fritsch Milling And Sizing Inc Powder Bottle 500 Ml, L-27
SDP
Catalog No. 501534081
Encompass
powder bottle 500 ml with screw lid, thread of sample bottle GL 55.
Can be used with dividing heads, division ratio 1:8 and 1:10.
Furthermore asample vessel 25 ml with lid, article-number 83.3190.16, is available, which can be inserted in the sample bottles 250 ml and 500 ml for dividing smallest sample quantities.
Catalog No. 50-153-4081 Supplier Fritsch Milling And Sizing Inc Supplier No. 27.1460.00
powder bottle 500 ml with screw lid, thread of sample bottle GL 55.
Can be used with dividing heads, division ratio 1:8 and 1:10.
Furthermore asample vessel 25 ml with lid, article-number 83.3190.16, is available, which can be inserted in the sample bottles 250 ml and 500 ml for dividing smallest sample quantities.