Learn More
Air Science Usa Powder Hood 48" Wide Balance Enclosure Pharmacy Hood
SDP
Catalog No. NC3521949
Encompass_Preferred
The Purair RX Balance Hood is a Class I enclosure that meets USP 800 requirements for non-sterile compounding procedures.
Catalog No. NC3521949 Supplier Air Science Usa Supplier No. P548XTRXA
The Purair RX Balance Hood is a Class I enclosure that meets USP 800 requirements for non-sterile compounding procedures.