Learn More
Braintree Scientific Dyets Powder Feeder Jar Setup,Complete Setup
SDP
Catalog No. NC0496158
Encompass
Dyets Powder Feeder Jar Setup. Prevents tipping or moving of Dyets feeding jar. 100% stainless steel for easy cleaning and durability. Versatile design allows for placement in any cage corner. Set contains jar, lid, follower, and holder. **Sold for research purposes only and not for human use**
Catalog No. NC0496158 Supplier Braintree Scientific Supplier No. DYETSSETUP
Dyets Powder Feeder Jar Setup. Prevents tipping or moving of Dyets feeding jar. 100% stainless steel for easy cleaning and durability. Versatile design allows for placement in any cage corner. Set contains jar, lid, follower, and holder. **Sold for research purposes only and not for human use**