Learn More
Prism Research Glass POWDER FUNNEL 14/20 JOINT
SDP
Catalog No. NC3302178
Encompass_Preferred
Powder funnel 14/20 joint offset straight side to be used with multi neck flask
Catalog No. NC3302178 Supplier Prism Research Glass Supplier No. PG926601
Add to Cart
Add to Cart
Powder funnel 14/20 joint offset straight side to be used with multi neck flask