Learn More
Chemglass Life Sciences 100mm Powder Funnel, 24/40 Inner Joint, 195mm OAL
SDP
Catalog No. CG1720L02
Encompass_Preferred
100mm Powder Funnel, 24/40 Inner Joint, 195mm OAL. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnel has steep sides to permit free-flow of solid materials. With standard taper inner joint.
Catalog No. CG1720L02 Supplier Chemglass Life Sciences Supplier No. CG1720L02
100mm Powder Funnel, 24/40 Inner Joint, 195mm OAL. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnel has steep sides to permit free-flow of solid materials. With standard taper inner joint.