Learn More
Ace Glass, Inc. Powder funnel, 75mm, angle heavy wall, 24/40 joint
SDP
Catalog No. 50979227
Encompass_Preferred
75mm heavy wall angle powder funnel with 24/40 standard taper joint. For use with pouring powders or liquids into multi-neck flasks.
Catalog No. 50-979-227 Supplier Ace Glass, Inc. Supplier No. 723806
75mm heavy wall angle powder funnel with 24/40 standard taper joint. For use with pouring powders or liquids into multi-neck flasks.