Learn More
Ace Glass, Inc. Powder funnel, 75mm, HW 24/40 joint
SDP
Catalog No. 50979223
Encompass_Preferred
75mm heavy wall powder funnel for greater durability when pouring powders or liquids into ground joint containers. Funnel has a 24/40 standard taper joint.
Catalog No. 50-979-223 Supplier Ace Glass, Inc. Supplier No. 723505
75mm heavy wall powder funnel for greater durability when pouring powders or liquids into ground joint containers. Funnel has a 24/40 standard taper joint.