Learn More
Analytical Sales & Services Powder Funnel Plate for 8x30mm Crimp Top Vials
Catalog No. 5000805105
Encompass
• Easily dispenses powder into your 8x30 Crimp Top Vials\n• Extra wide openings for easy filling\n• Funneled holes guide powder down into vials\n• Circular grooves on underside of transfer plate “lock” vials into position to eliminate spillage and mess\n• Can be used with 96-Well Stackable Trays with 8x30mm Crimp Top Vials (884008)
Catalog No. 50-008-05105 Supplier Analytical Sales & Services Supplier No. 96T102
• Easily dispenses powder into your 8x30 Crimp Top Vials\n• Extra wide openings for easy filling\n• Funneled holes guide powder down into vials\n• Circular grooves on underside of transfer plate “lock” vials into position to eliminate spillage and mess\n• Can be used with 96-Well Stackable Trays with 8x30mm Crimp Top Vials (884008)