Learn More
BURKLE INC Disposable powder funnel, PS, Ø 3.94 in (100 mm), white, sterile
Catalog No. 503081208
Encompass_Preferred
The disposable powder funnel has a short and wide outlet and is suitable for transferring powdered samples and materials, granulates, suspensions and other liquids.
The wide opening of the disposable powder funnel prevents the powder or granulate from clogging during filling. The practical and hygienic filling aid is therefore a useful accessory when taking samples or in the laboratory.
In combination with a filter paper, the disposable funnel is also very suitable for filtering.
Catalog No. 50-308-1208 Supplier BURKLE INC Supplier No. 53786006
The disposable powder funnel has a short and wide outlet and is suitable for transferring powdered samples and materials, granulates, suspensions and other liquids.
The wide opening of the disposable powder funnel prevents the powder or granulate from clogging during filling. The practical and hygienic filling aid is therefore a useful accessory when taking samples or in the laboratory.
In combination with a filter paper, the disposable funnel is also very suitable for filtering.