Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Powder Packs™: Silica HR Citric Acid

Catalog No. 09054100 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
100 Pack
Range:
0 to 1.6mg/L
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Range Volume (Metric)
09-054-100 100 Pack 0 to 1.6mg/L 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-054-100 Supplier Lovibond™ Supplier No. 531710
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pre-measured, unit dose reagents for water quality testing

  • Simply tear open Powder Pack™ and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open the package
  • Easily create a “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in the corners

Specifications

Color White
Composition Citric Acid
pH Range 2.2
Volume (Metric) 10 mL
Format Powder
For Use With (Application) Water quality testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
Quantity 100 Pack
Range 0 to 1.6mg/L
Storage Requirements 20°C
Type Silica HR Citric Acid Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.