Learn More
Perkin Elmer US LLC Powder Sample Holder for the Front Surface Accessory
Catalog No. 502113288
Encompass_Preferred
The Front Surface Accessory broadens the range of samples available for fluorescence and phosphorescence measurements from solid and semi-solid samples to opaque and turbid samples. This accessory can be adjusted to optimize sensitivity and is compatible with the LS 45/50/50B/55 spectrometers.
Catalog No. 50-211-3288 Supplier Perkin Elmer US LLC Supplier No. 52123164
The Front Surface Accessory broadens the range of samples available for fluorescence and phosphorescence measurements from solid and semi-solid samples to opaque and turbid samples. This accessory can be adjusted to optimize sensitivity and is compatible with the LS 45/50/50B/55 spectrometers.